2-(1,4-Diazepan-1-yl)[1,3]oxazolo[5,4-b]pyridine

Supplier: Matrix Scientific

035948-500MG
101840-388EA 484.62 USD
101840-388
2-(1,4-Diazepan-1-yl)[1,3]oxazolo[5,4-b]pyridine
2-(1,4-Diazepan-1-yl)[1,3]oxazolo[5,4-b]pyridine

Hazard: Irritant

Formula: C₁₁H₁₄N₄O
MDL Number: MFCD11696403
CAS Number: 1071369-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR