Ethyl 2-(piperidin-3-yl)acetate hydrochloride

Supplier: Matrix Scientific

Synonyms: Ethyl piperidin-3-ylacetate hydrochloride

035947-500MG
101840-386EA 289.8 USD
101840-386
Ethyl 2-(piperidin-3-yl)acetate hydrochloride
Ethyl 2-(piperidin-3-yl)acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₂
MW: 207.7 g/mol
MDL Number: MFCD08059314
CAS Number: 64995-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR