2-(1,4-Diazepan-1-yl)-5-methyl[1,3]oxazolo[4,5-b]pyridine

Supplier: Matrix Scientific

035946-500MG
101840-384EA 484.62 USD
101840-384
2-(1,4-Diazepan-1-yl)-5-methyl[1,3]oxazolo[4,5-b]pyridine
2-(1,4-Diazepan-1-yl)-5-methyl[1,3]oxazolo[4,5-b]pyridine

Hazard: Irritant

Formula: C₁₂H₁₆N₄O
MDL Number: MFCD09701646
CAS Number: 1035840-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR