2-Piperazin-1-yl[1,3]oxazolo[4,5-b]pyridine

Supplier: Matrix Scientific

035945-500MG
101840-382EA 484.62 USD
101840-382
2-Piperazin-1-yl[1,3]oxazolo[4,5-b]pyridine
2-Piperazin-1-yl[1,3]oxazolo[4,5-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MDL Number: MFCD09743365
CAS Number: 113520-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR