1-(Methoxyacetyl)-1,4-diazepane hydrochloride

Supplier: Matrix Scientific

035941-500MG
101840-374EA 290.77 USD
101840-374
1-(Methoxyacetyl)-1,4-diazepane hydrochloride
1-(Methoxyacetyl)-1,4-diazepane hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O₂
MDL Number: MFCD11696399
CAS Number: 926191-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR