2-Chloro-N-ethyl-N-methylacetamide

Supplier: Matrix Scientific

035939-500MG
101840-370EA 11.5 USD
101840-370
2-Chloro-N-ethyl-N-methylacetamide
2-Chloro-N-ethyl-N-methylacetamide

Hazard: Irritant

Formula: C₅H₁₀ClNO
Boiling Pt: 64…65 °C (0.7 Torr)
MDL Number: MFCD10686581
CAS Number: 2746-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR