(5-Methoxy-1,3-benzoxazol-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

035935-500MG
101840-362EA 402.5 USD
101840-362
(5-Methoxy-1,3-benzoxazol-2-yl)methylamine hydrochloride
(5-Methoxy-1,3-benzoxazol-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD11696398
CAS Number: 944897-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR