1-(Pyridin-2-ylcarbonyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

035930-500MG
101840-352EA 193.85 USD
101840-352
1-(Pyridin-2-ylcarbonyl)piperidine-4-carboxylic acid
1-(Pyridin-2-ylcarbonyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
MDL Number: MFCD08442613
CAS Number: 67691-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR