1-Propionylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

035928-500MG
101840-348EA 338.46 USD
101840-348
1-Propionylpyrrolidine-2-carboxylic acid
1-Propionylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD09049909
CAS Number: 59785-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR