1-Isonicotinoylpiperidine-4-carboxylic acid

Supplier: Matrix Scientific

035926-500MG
101840-344EA 144.9 USD
101840-344
1-Isonicotinoylpiperidine-4-carboxylic acid
1-Isonicotinoylpiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
MDL Number: MFCD08442824
CAS Number: 67691-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR