Methyl-1H-imidazol-1-ylacetate

Supplier: Matrix Scientific

035925-500MG
101840-342EA 24.62 USD
101840-342
Methyl-1H-imidazol-1-ylacetate
Methyl-1H-imidazol-1-ylacetate

Hazard: Irritant/Refrigerate

Formula: C₆H₈N₂O₂
MDL Number: MFCD11183411
CAS Number: 25023-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR