Ethyl pyrazine-2-carboxylate
Supplier: Matrix Scientific
035922-500MG
101840-336EA
20
USD
101840-336
Ethyl pyrazine-2-carboxylate
Ethyl pyrazine-2-carboxylate
Hazard: Irritant
Formula:
C₇H₈N₂O₂ |
MDL Number:
MFCD00792887 CAS Number: 6924-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...