(5-tert-Butyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

035921-500MG
101840-334EA 193.85 USD
101840-334
(5-tert-Butyl-1,3-benzoxazol-2-yl)methylamine hydrochloride
(5-tert-Butyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD11696396
CAS Number: 1119449-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR