1-(Thien-2-ylcarbonyl)piperidin-4-one

Supplier: Matrix Scientific

035920-500MG
101840-332EA 109.25 USD
101840-332
1-(Thien-2-ylcarbonyl)piperidin-4-one
1-(Thien-2-ylcarbonyl)piperidin-4-one

Hazard: Irritant

Formula: C₁₀H₁₁NO₂S
MDL Number: MFCD09939609
CAS Number: 141945-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR