1-(Cyclobutylcarbonyl)-1,4-diazepane

Supplier: Matrix Scientific

035917-500MG
101840-326EA 362.25 USD
101840-326
1-(Cyclobutylcarbonyl)-1,4-diazepane
1-(Cyclobutylcarbonyl)-1,4-diazepane

Hazard: Irritant

Formula: C₁₀H₁₈N₂O
MDL Number: MFCD09042773
CAS Number: 926193-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR