1-(3-Methylbutanoyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

035915-500MG
101840-322EA 146.15 USD
101840-322
1-(3-Methylbutanoyl)piperidine-4-carboxylic acid
1-(3-Methylbutanoyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD08442413
CAS Number: 926238-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR