1-(Cyclopropylcarbonyl)piperidin-4-one

Supplier: Matrix Scientific

035913-500MG
101840-318EA 217.35 USD
101840-318
1-(Cyclopropylcarbonyl)piperidin-4-one
1-(Cyclopropylcarbonyl)piperidin-4-one

Hazard: Irritant

Formula: C₉H₁₃NO₂
MDL Number: MFCD09949954
CAS Number: 63463-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR