1-Butyrylpiperidin-4-one

Supplier: Matrix Scientific

035912-500MG
101840-316EA 144.9 USD
101840-316
1-Butyrylpiperidin-4-one
1-Butyrylpiperidin-4-one

Hazard: Irritant

Formula: C₉H₁₅NO₂
MDL Number: MFCD09864589
CAS Number: 959241-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR