N-(2-Aminoethyl)-2,2,2-trifluoroacetamide hydrochloride

Supplier: Matrix Scientific

035911-500MG
101840-314EA 316.25 USD
101840-314
N-(2-Aminoethyl)-2,2,2-trifluoroacetamide hydrochloride
N-(2-Aminoethyl)-2,2,2-trifluoroacetamide hydrochloride

Hazard: Irritant

Formula: C₄H₈ClF₃N₂O
MDL Number: MFCD01630946
CAS Number: 5458-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR