1-(Cyclobutylcarbonyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

035908-500MG
101840-308EA 217.35 USD
101840-308
1-(Cyclobutylcarbonyl)piperidine-4-carboxylic acid
1-(Cyclobutylcarbonyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD02679040
CAS Number: 700815-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR