1-Isobutyrylpiperidin-4-one

Supplier: Matrix Scientific

035907-500MG
101840-306EA 144.9 USD
101840-306
1-Isobutyrylpiperidin-4-one
1-Isobutyrylpiperidin-4-one

Hazard: Irritant

Formula: C₉H₁₅NO₂
MDL Number: MFCD09701667
CAS Number: 86996-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR