2-[4-(1,3-Benzoxazol-2-yl)morpholin-2-yl]ethanamine hydrochloride
Supplier: Matrix Scientific
035904-500MG
101840-300EA
484.62
USD
101840-300
2-[4-(1,3-Benzoxazol-2-yl)morpholin-2-yl]ethanamine hydrochloride
2-[4-(1,3-Benzoxazol-2-yl)morpholin-2-yl]ethanamine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClN₃O₂ |
MDL Number:
MFCD11696390 CAS Number: 1158772-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...