(1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpyrrolidin-3-yl)methylamine dihydrochloride

Supplier: Matrix Scientific

035903-500MG
101840-298EA 362.25 USD
101840-298
(1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpyrrolidin-3-yl)methylamine dihydrochloride
(1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpyrrolidin-3-yl)methylamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆Cl₂N₄O
MDL Number: MFCD11696389
CAS Number: 1035840-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR