(1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpiperidin-3-yl)acetic acid

Supplier: Matrix Scientific

035902-500MG
101840-296EA 362.25 USD
101840-296
(1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpiperidin-3-yl)acetic acid
(1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpiperidin-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₃
MDL Number: MFCD09701631
CAS Number: 1035840-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR