2-(4-[1,3]Oxazolo[4,5-b]pyridin-2-ylmorpholin-2-yl)ethanamine dihydrochloride

Supplier: Matrix Scientific

035898-500MG
101840-288EA 484.62 USD
101840-288
2-(4-[1,3]Oxazolo[4,5-b]pyridin-2-ylmorpholin-2-yl)ethanamine dihydrochloride
2-(4-[1,3]Oxazolo[4,5-b]pyridin-2-ylmorpholin-2-yl)ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈Cl₂N₄O₂
MDL Number: MFCD11696388
CAS Number: 1158736-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR