6-Chloro-2-(chloromethyl)-1,3-benzoxazole

Supplier: Matrix Scientific

035895-500MG
101840-282EA 218.46 USD
101840-282
6-Chloro-2-(chloromethyl)-1,3-benzoxazole
6-Chloro-2-(chloromethyl)-1,3-benzoxazole

Hazard: Irritant

Formula: C₈H₅Cl₂NO
MDL Number: MFCD09261477
CAS Number: 202396-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR