(6-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

035893-500MG
101840-278EA 387.69 USD
101840-278
(6-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride
(6-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MDL Number: MFCD11696385
CAS Number: 1158477-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR