[1-(6-Chloro-1,3-benzoxazol-2-yl)piperidin-3-yl]acetic acid

Supplier: Matrix Scientific

035890-500MG
101840-272EA 484.62 USD
101840-272
[1-(6-Chloro-1,3-benzoxazol-2-yl)piperidin-3-yl]acetic acid
[1-(6-Chloro-1,3-benzoxazol-2-yl)piperidin-3-yl]acetic acid

Hazard: Irritant

Formula: C₁₄H₁₅ClN₂O₃
MDL Number: MFCD09701663
CAS Number: 1035840-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR