6-Methyl-2-piperidin-3-yl-1,3-benzoxazole hydrochloride

Supplier: Matrix Scientific

035889-500MG
101840-270EA 289.8 USD
101840-270
6-Methyl-2-piperidin-3-yl-1,3-benzoxazole hydrochloride
6-Methyl-2-piperidin-3-yl-1,3-benzoxazole hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O
MDL Number: MFCD11696384
CAS Number: 1158757-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR