4-Hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

035881-500MG
101840-254EA 174.8 USD
101840-254
4-Hydrazino-4-oxobutanamide
4-Hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₄H₉N₃O₂
MDL Number: MFCD00984930
CAS Number: 130673-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR