1-(5-tert-Butyl-1,3-benzoxazol-2-yl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

035878-500MG
101840-248EA 484.62 USD
101840-248
1-(5-tert-Butyl-1,3-benzoxazol-2-yl)piperidine-4-carboxylic acid
1-(5-tert-Butyl-1,3-benzoxazol-2-yl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₃
MDL Number: MFCD09701680
CAS Number: 1035841-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR