1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpiperidine-4-carboxylic acid

Supplier: Matrix Scientific

035876-500MG
101840-244EA 484.62 USD
101840-244
1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpiperidine-4-carboxylic acid
1-[1,3]Oxazolo[4,5-b]pyridin-2-ylpiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₃
MDL Number: MFCD09701681
CAS Number: 936074-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR