(4-[1,3]Oxazolo[4,5-b]pyridin-2-ylmorpholin-2-yl)methylamine dihydrochloride

Supplier: Matrix Scientific

035867-500MG
101840-226EA 484.62 USD
101840-226
(4-[1,3]Oxazolo[4,5-b]pyridin-2-ylmorpholin-2-yl)methylamine dihydrochloride
(4-[1,3]Oxazolo[4,5-b]pyridin-2-ylmorpholin-2-yl)methylamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆Cl₂N₄O₂
MDL Number: MFCD11696374
CAS Number: 1158406-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR