4-(2-Acetylhydrazino)-4-oxobutanoic acid

Supplier: Matrix Scientific

035862-500MG
101840-216EA 290.77 USD
101840-216
4-(2-Acetylhydrazino)-4-oxobutanoic acid
4-(2-Acetylhydrazino)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₆H₁₀N₂O₄
MDL Number: MFCD11696373
CAS Number: 257885-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR