(5-Ethyl-1,3,4-oxadiazol-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

035861-500MG
101840-214EA 387.69 USD
101840-214
(5-Ethyl-1,3,4-oxadiazol-2-yl)methylamine hydrochloride
(5-Ethyl-1,3,4-oxadiazol-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₃O
MDL Number: MFCD11696372
CAS Number: 944897-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR