5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid
Supplier: Matrix Scientific
035860-500MG
101840-212EA
484.62
USD
101840-212
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid
Hazard: Irritant
Formula:
C₇H₉N₃O₂ |
MDL Number:
MFCD09701670 CAS Number: 1035841-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...