4-Hydrazino-N-(3-methylphenyl)-4-oxobutanamide

Supplier: Matrix Scientific

035859-500MG
101840-210EA 290.77 USD
101840-210
4-Hydrazino-N-(3-methylphenyl)-4-oxobutanamide
4-Hydrazino-N-(3-methylphenyl)-4-oxobutanamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD02255634
CAS Number: 443864-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR