2-[2-(5,7-Dichloro-1,3-benzoxazol-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

035857-500MG
101840-206EA 193.85 USD
101840-206
2-[2-(5,7-Dichloro-1,3-benzoxazol-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione
2-[2-(5,7-Dichloro-1,3-benzoxazol-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₇H₁₀Cl₂N₂O₃
MDL Number: MFCD11696370
CAS Number: 1071369-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR