2-(Chloromethyl)-4,5-dimethyl-1H-benzimidazole

Supplier: Matrix Scientific

035849-500MG
101840-190EA 116.92 USD
101840-190
2-(Chloromethyl)-4,5-dimethyl-1H-benzimidazole
2-(Chloromethyl)-4,5-dimethyl-1H-benzimidazole

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂
MDL Number: MFCD09701678
CAS Number: 405173-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR