(5-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

035846-500MG
101840-184EA 461.54 USD
101840-184
(5-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride
(5-Methyl-1,3-benzoxazol-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MDL Number: MFCD11696367
CAS Number: 944897-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR