tert-Butyl-2-morpholin-2-ylethylcarbamate

Supplier: Matrix Scientific

035844-500MG
101840-180EA 338.46 USD
101840-180
tert-Butyl-2-morpholin-2-ylethylcarbamate
tert-Butyl-2-morpholin-2-ylethylcarbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₃
MW: 230.31 g/mol
MDL Number: MFCD09701685
CAS Number: 1032507-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR