Ethyl-3-chloropropanimidoate hydrochloride

Supplier: Matrix Scientific

035839-500MG
101840-170EA 193.85 USD
101840-170
Ethyl-3-chloropropanimidoate hydrochloride
Ethyl-3-chloropropanimidoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₁Cl₂NO
MDL Number: MFCD11696357
CAS Number: 21367-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR