Ethyl-4-chlorobutanimidoate hydrochloride

Supplier: Matrix Scientific

035838-500MG
101840-168EA 193.85 USD
101840-168
Ethyl-4-chlorobutanimidoate hydrochloride
Ethyl-4-chlorobutanimidoate hydrochloride

Hazard: Irritant

Formula: C₆H₁₃Cl₂NO
MDL Number: MFCD11696356
CAS Number: 89224-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR