1-(2-Methylpiperidin-1-yl)acetone

Supplier: Matrix Scientific

035836-500MG
101840-164EA 146.15 USD
101840-164
1-(2-Methylpiperidin-1-yl)acetone
1-(2-Methylpiperidin-1-yl)acetone

Hazard: Irritant

Formula: C₉H₁₇NO
MDL Number: MFCD09864601
CAS Number: 24633-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR