1-Azepan-1-ylacetone hydrochloride

Supplier: Matrix Scientific

035833-500MG
101840-158EA 193.85 USD
101840-158
1-Azepan-1-ylacetone hydrochloride
1-Azepan-1-ylacetone hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO
MDL Number: MFCD09864566
CAS Number: 1158643-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR