1-[Ethyl(methyl)amino]acetone hydrochloride

Supplier: Matrix Scientific

035832-500MG
101840-156EA 387.69 USD
101840-156
1-[Ethyl(methyl)amino]acetone hydrochloride
1-[Ethyl(methyl)amino]acetone hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MDL Number: MFCD11696355
CAS Number: 1158315-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR