1-(2-Ethoxyphenyl)ethanamine

Supplier: Matrix Scientific

035828-500MG
101840-150EA 484.62 USD
101840-150
1-(2-Ethoxyphenyl)ethanamine
1-(2-Ethoxyphenyl)ethanamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD05215234
CAS Number: 603945-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR