2-(1,4-Diazepan-1-yl)-1,3-benzothiazoletrifluoroacetic acid salt

Supplier: Matrix Scientific

035827-500MG
101840-148EA 290.77 USD
101840-148
2-(1,4-Diazepan-1-yl)-1,3-benzothiazoletrifluoroacetic acid salt
2-(1,4-Diazepan-1-yl)-1,3-benzothiazoletrifluoroacetic acid salt

Hazard: Irritant

Formula: C₁₄H₁₆F₃N₃O₂S
MDL Number: MFCD12026774
CAS Number: 1185033-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR