N-(2-Methoxyethyl)-N-(tetrahydrofuran-2-ylmethyl)amine

Supplier: Matrix Scientific

035826-500MG
101840-146EA 146.15 USD
101840-146
N-(2-Methoxyethyl)-N-(tetrahydrofuran-2-ylmethyl)amine
N-(2-Methoxyethyl)-N-(tetrahydrofuran-2-ylmethyl)amine

Hazard: Irritant

Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
MDL Number: MFCD08059959
CAS Number: 55100-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR