Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phenylglycine

Supplier: Matrix Scientific


035817-500MG
101840-126EA 164.77 USD InStock
101840-126
N-Phenylglycine
N-Phenylglycine
CAS: 103-01-5
MDL Number: MFCD00014009
Molecular Formula: C8H9NO2
Molecular Weight: 151,17
Hazard: Irritant


Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty